Imprimer

Anti-AURKC Mouse Monoclonal Antibody [clone: OTI2B7]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500602EA 608 EUR
ORIGTA500602
Anti-AURKC Mouse Monoclonal Antibody [clone: OTI2B7]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: AURKC (aurora kinase C)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR