Imprimer

Anti-POR Mouse Monoclonal Antibody [clone: OTI5B3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500612EA 603 EUR
ORIGTA500612
Anti-POR Mouse Monoclonal Antibody [clone: OTI5B3]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:50; Flow Cytometry: 1:100

Type: Primary
Antigen: POR (P450 (cytochrome) oxidoreductase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR