Imprimer

Anti-MAD2L1 Mouse Monoclonal Antibody [clone: OTI4D2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500617EA 608 EUR
ORIGTA500617
Anti-MAD2L1 Mouse Monoclonal Antibody [clone: OTI4D2]
Anticorps
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: MAD2L1 (MAD2 mitotic arrest deficient-like 1 (yeast))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR