Imprimer

Anti-CA9 Mouse monoclonal antibody [clone: OTI1G7]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500623EA 608 EUR
ORIGTA500623
Anti-CA9 Mouse monoclonal antibody [clone: OTI1G7]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Flow Cytometry: 1:100

Type: Primary
Antigen: CA9 (carbonic anhydrase IX)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR