Imprimer

Anti-PDE10A Mouse Monoclonal Antibody [clone: OTI1C9]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500625EA 603 EUR
ORIGTA500625
Anti-PDE10A Mouse Monoclonal Antibody [clone: OTI1C9]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50

Type: Primary
Antigen: PDE10A (phosphodiesterase 10A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR