Imprimer

Anti-FCGR2A Mouse Monoclonal Antibody [clone: OTI6D11]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500653EA 603 EUR
ORIGTA500653
Anti-FCGR2A Mouse Monoclonal Antibody [clone: OTI6D11]
Anticorps
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:50; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: FCGR2A (Fc fragment of IgG, low affinity IIa, receptor (CD32))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR