Imprimer

Anti-GBP2 Mouse Monoclonal Antibody [clone: OTI3B8]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500656EA 581 EUR
ORIGTA500656
Anti-GBP2 Mouse Monoclonal Antibody [clone: OTI3B8]
Anticorps
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: GBP2 (guanylate binding protein 2, interferon-inducible)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR