Imprimer
Anti-LTA4H Mouse Monoclonal Antibody [clone: OTI6B5]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA500663EA 581 EUR
ORIGTA500663
Anti-LTA4H Mouse Monoclonal Antibody [clone: OTI6B5]
Anticorps
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Flow Cytometry: 1:100; Immunoprecipitation: 2-4μg/mg
Type: Primary
Antigen: LTA4H (leukotriene A4 hydrolase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human