Imprimer

Anti-LTA4H Mouse Monoclonal Antibody [clone: OTI1E9]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500666EA 586 EUR
ORIGTA500666
Anti-LTA4H Mouse Monoclonal Antibody [clone: OTI1E9]
Anticorps
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunoprecipitation: 2-4μg/mg

Type: Primary
Antigen: LTA4H (leukotriene A4 hydrolase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR