Imprimer

Anti-RNF144B Mouse Monoclonal Antibody [clone: OTI12B6]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500709EA 603 EUR
ORIGTA500709
Anti-RNF144B Mouse Monoclonal Antibody [clone: OTI12B6]
Anticorps
Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: RNF144B (ring finger protein 144B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR