Imprimer

Anti-IFIT3 Mouse Monoclonal Antibody [clone: OTI1G1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500726EA 608 EUR
ORIGTA500726
Anti-IFIT3 Mouse Monoclonal Antibody [clone: OTI1G1]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: IFIT3 (interferon-induced protein with tetratricopeptide repeats 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR