Imprimer
Anti-AKR1A1 Mouse Monoclonal Antibody [clone: OTI8E12]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA500741EA 603 EUR
ORIGTA500741
Anti-AKR1A1 Mouse Monoclonal Antibody [clone: OTI8E12]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Flow Cytometry: 1:100
Type: Primary
Antigen: AKR1A1 (aldo-keto reductase family 1, member A1 (aldehyde reductase))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human