Imprimer
Anti-SORD Mouse Monoclonal Antibody [clone: OTI9G9]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA500785EA 603 EUR
ORIGTA500785
Anti-SORD Mouse Monoclonal Antibody [clone: OTI9G9]
Anticorps
Recommended Dilutions: Immunofluorescence: 1:100 Flow Cytometry: 1:100
Type: Primary
Antigen: SORD (sorbitol dehydrogenase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human