Imprimer

Anti-RPA2 Mouse Monoclonal Antibody [clone: OTI8B3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500788EA 603 EUR
ORIGTA500788
Anti-RPA2 Mouse Monoclonal Antibody [clone: OTI8B3]
Anticorps
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: RPA2 (replication protein A2, 32kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR