Imprimer

Anti-ID3 Mouse Monoclonal Antibody [clone: OTI12C7]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500790EA 603 EUR
ORIGTA500790
Anti-ID3 Mouse Monoclonal Antibody [clone: OTI12C7]
Anticorps
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ID3 (inhibitor of DNA binding 3, dominant negative helix-loop-helix protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR