Imprimer

Anti-TTLL12 Mouse Monoclonal Antibody [clone: OTI12B7]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500806EA 581 EUR
ORIGTA500806
Anti-TTLL12 Mouse Monoclonal Antibody [clone: OTI12B7]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: TTLL12 (tubulin tyrosine ligase-like family, member 12)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR