Imprimer

Anti-AKT2 Mouse Monoclonal Antibody [clone: OTI4G1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500831EA 603 EUR
ORIGTA500831
Anti-AKT2 Mouse Monoclonal Antibody [clone: OTI4G1]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: AKT2 (v-akt murine thymoma viral oncogene homolog 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR