Imprimer
Anti-PLEK Mouse Monoclonal Antibody [clone: OTI4F10]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA500899EA 581 EUR
ORIGTA500899
Anti-PLEK Mouse Monoclonal Antibody [clone: OTI4F10]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Flow Cytometry: 1:100
Type: Primary
Antigen: PLEK (pleckstrin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human