Imprimer

Anti-PSMC3 Mouse Monoclonal Antibody [clone: OTI7D1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500930EA 581 EUR
ORIGTA500930
Anti-PSMC3 Mouse Monoclonal Antibody [clone: OTI7D1]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: PSMC3 (proteasome (prosome, macropain) 26S subunit, ATPase, 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR