Imprimer
Anti-TPMT Mouse Monoclonal Antibody [clone: OTI2A2]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA500966EA 581 EUR
ORIGTA500966
Anti-TPMT Mouse Monoclonal Antibody [clone: OTI2A2]
Anticorps
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100
Type: Primary
Antigen: TPMT (thiopurine S-methyltransferase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human