Imprimer

Anti-LIPG Mouse Monoclonal Antibody [clone: OTI1D9]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501016EA 603 EUR
ORIGTA501016
Anti-LIPG Mouse Monoclonal Antibody [clone: OTI1D9]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: LIPG (lipase, endothelial)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR