Imprimer

Anti-PTPRE Mouse Monoclonal Antibody [clone: OTI2C11]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501025EA 603 EUR
ORIGTA501025
Anti-PTPRE Mouse Monoclonal Antibody [clone: OTI2C11]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: PTPRE (protein tyrosine phosphatase, receptor type, E)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR