Imprimer

Anti-PTPRE Mouse Monoclonal Antibody [clone: OTI4B2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501028EA 603 EUR
ORIGTA501028
Anti-PTPRE Mouse Monoclonal Antibody [clone: OTI4B2]
Anticorps
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:50

Type: Primary
Antigen: PTPRE (protein tyrosine phosphatase, receptor type, E)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR