Imprimer

Anti-NPR3 Mouse Monoclonal Antibody [clone: OTI11A6]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501076EA 603 EUR
ORIGTA501076
Anti-NPR3 Mouse Monoclonal Antibody [clone: OTI11A6]
Anticorps
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: NPR3 (natriuretic peptide receptor 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR