Imprimer

Anti-NME4 Mouse Monoclonal Antibody [clone: OTI1A5]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA501110
ORIGTA501110EA 616 EUR
ORIGTA501110
Anti-NME4 Mouse Monoclonal Antibody [clone: OTI1A5]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: NME4 (NME/NM23 nucleoside diphosphate kinase 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR