Imprimer

Anti-CAPZA1 Mouse Monoclonal Antibody [clone: OTI2G4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501124EA 616 EUR
ORIGTA501124
Anti-CAPZA1 Mouse Monoclonal Antibody [clone: OTI2G4]
Anticorps
  • Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50 Flow Cytometry: 1:100

Type: Primary
Antigen: CAPZA1 (capping protein (actin filament) muscle Z-line, alpha 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR