Imprimer

Anti-ALDH3A1 Mouse Monoclonal Antibody [clone: OTI1C6]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501139EA 603 EUR
ORIGTA501139
Anti-ALDH3A1 Mouse Monoclonal Antibody [clone: OTI1C6]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: ALDH3A1 (aldehyde dehydrogenase 3 family, member A1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR