Imprimer

Anti-ERCC1 Mouse Monoclonal Antibody [clone: OTI1B3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501167EA 603 EUR
ORIGTA501167
Anti-ERCC1 Mouse Monoclonal Antibody [clone: OTI1B3]
Anticorps
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:50-1:100

Type: Primary
Antigen: ERCC1 (excision repair cross-complementation group 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR