Imprimer
Anti-BSG Mouse Monoclonal Antibody [clone: OTI10E10]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA501189EA 603 EUR
ORIGTA501189
Anti-BSG Mouse Monoclonal Antibody [clone: OTI10E10]
Anticorps
  • Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: BSG (basigin (Ok blood group))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant