Imprimer

Anti-ACAT2 Mouse Monoclonal Antibody [clone: OTI5F7]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501219EA 603 EUR
ORIGTA501219
Anti-ACAT2 Mouse Monoclonal Antibody [clone: OTI5F7]
Anticorps
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ACAT2 (acetyl-CoA acetyltransferase 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR