Imprimer

Anti-ACAT2 Mouse Monoclonal Antibody [clone: OTI7B1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA501221
ORIGTA501221EA 616 EUR
ORIGTA501221
Anti-ACAT2 Mouse Monoclonal Antibody [clone: OTI7B1]
Anticorps
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ACAT2 (acetyl-CoA acetyltransferase 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR