Imprimer

Anti-DDX39B Mouse Monoclonal Antibody [clone: OTI2E4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501235EA 608 EUR
ORIGTA501235
Anti-DDX39B Mouse Monoclonal Antibody [clone: OTI2E4]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: DDX39B (DEAD (Asp-Glu-Ala-Asp) box polypeptide 39B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR