Imprimer

Anti-CYP1A2 Mouse Monoclonal Antibody [clone: OTI2C6]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501244EA 603 EUR
ORIGTA501244
Anti-CYP1A2 Mouse Monoclonal Antibody [clone: OTI2C6]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: CYP1A2 (cytochrome P450, family 1, subfamily A, polypeptide 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR