Imprimer

Anti-GNMT Mouse Monoclonal Antibody [clone: OTI8A3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501276EA 586 EUR
ORIGTA501276
Anti-GNMT Mouse Monoclonal Antibody [clone: OTI8A3]
Anticorps
Recommended Dilutions: Western Blot: 1:500 Flow Cytometry: 1:100

Type: Primary
Antigen: GNMT (glycine N-methyltransferase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR