Imprimer

Anti-FHL1 Mouse Monoclonal Antibody [clone: OTI2E1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501280EA 603 EUR
ORIGTA501280
Anti-FHL1 Mouse Monoclonal Antibody [clone: OTI2E1]
Anticorps
Recommended Dilutions: Immunofluorescence: 1:100

Type: Primary
Antigen: FHL1 (four and a half LIM domains 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR