Imprimer

Anti-USP5 Mouse Monoclonal Antibody [clone: OTI1F9]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501297EA 581 EUR
ORIGTA501297
Anti-USP5 Mouse Monoclonal Antibody [clone: OTI1F9]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: USP5 (ubiquitin specific peptidase 5 (isopeptidase T))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR