Imprimer

Anti-HIBCH Mouse Monoclonal Antibody [clone: OTI1C7]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501319EA 581 EUR
ORIGTA501319
Anti-HIBCH Mouse Monoclonal Antibody [clone: OTI1C7]
Anticorps
  • Recommended Dilutions: Western Blot: 1:1000-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HIBCH (3-hydroxyisobutyryl-CoA hydrolase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR