Imprimer
Anti-SNX9 Mouse Monoclonal Antibody [clone: OTI2A11]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA501323EA 603 EUR
ORIGTA501323
Anti-SNX9 Mouse Monoclonal Antibody [clone: OTI2A11]
Anticorps
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: SNX9 (sorting nexin 9)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant