Imprimer
Anti-TBXAS1 Mouse Monoclonal Antibody [clone: OTI1C3]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA501379EA 603 EUR
ORIGTA501379
Anti-TBXAS1 Mouse Monoclonal Antibody [clone: OTI1C3]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: TBXAS1 (thromboxane A synthase 1 (platelet))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human