Imprimer

Anti-COX6A1 Mouse Monoclonal Antibody [clone: OTI3F1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501397EA 603 EUR
ORIGTA501397
Anti-COX6A1 Mouse Monoclonal Antibody [clone: OTI3F1]
Anticorps
Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: COX6A1 (cytochrome c oxidase subunit VIa polypeptide 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR