Imprimer

Anti-C15orf40 Mouse Monoclonal Antibody [clone: OTI2B7]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501400EA 608 EUR
ORIGTA501400
Anti-C15orf40 Mouse Monoclonal Antibody [clone: OTI2B7]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Flow Cytometry: 1:100

Type: Primary
Antigen: C15orf40 (chromosome 15 open reading frame 40)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR