Imprimer

Anti-C10orf82 Mouse Monoclonal Antibody [clone: OTI1G4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501481EA 608 EUR
ORIGTA501481
Anti-C10orf82 Mouse Monoclonal Antibody [clone: OTI1G4]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: C10orf82 (chromosome 10 open reading frame 82)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR