Imprimer

Anti-SHPRH Mouse Monoclonal Antibody [clone: OTI7B2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501497EA 581 EUR
ORIGTA501497
Anti-SHPRH Mouse Monoclonal Antibody [clone: OTI7B2]
Anticorps
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SHPRH (SNF2 histone linker PHD RING helicase, E3 ubiquitin protein ligase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR