Imprimer

Anti-QPRT Mouse Monoclonal Antibody [clone: OTI2H4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501526EA 603 EUR
ORIGTA501526
Anti-QPRT Mouse Monoclonal Antibody [clone: OTI2H4]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:50

Type: Primary
Antigen: QPRT (quinolinate phosphoribosyltransferase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR