Imprimer
Anti-BTN1A1 Mouse Monoclonal Antibody [clone: OTI4H10]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA501532EA 603 EUR
ORIGTA501532
Anti-BTN1A1 Mouse Monoclonal Antibody [clone: OTI4H10]
Anticorps
  • Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100
Type: Primary
Antigen: BTN1A1 (butyrophilin, subfamily 1, member A1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human