Imprimer
Anti-CD80 Mouse Monoclonal Antibody [clone: OTI2H1]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA501577EA 603 EUR
ORIGTA501577
Anti-CD80 Mouse Monoclonal Antibody [clone: OTI2H1]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: CD80 (CD80 molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human