Imprimer

Anti-NUMB Mouse Monoclonal Antibody [clone: OTI6B2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA501613
ORIGTA501613EA 616 EUR
ORIGTA501613
Anti-NUMB Mouse Monoclonal Antibody [clone: OTI6B2]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: NUMB (numb homolog (Drosophila))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR