Imprimer

Anti-SULT2A1 Mouse Monoclonal Antibody [clone: OTI3G4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501656EA 608 EUR
ORIGTA501656
Anti-SULT2A1 Mouse Monoclonal Antibody [clone: OTI3G4]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: SULT2A1 (sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone (DHEA)-preferring, member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR