Imprimer

Anti-C16orf72 Mouse Monoclonal Antibody [clone: OTI3E5]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501673EA 581 EUR
ORIGTA501673
Anti-C16orf72 Mouse Monoclonal Antibody [clone: OTI3E5]
Anticorps
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: C16orf72 (chromosome 16 open reading frame 72)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR