Imprimer

Anti-TUBA3E Mouse Monoclonal Antibody [clone: OTI10B4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501715EA 603 EUR
ORIGTA501715
Anti-TUBA3E Mouse Monoclonal Antibody [clone: OTI10B4]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: TUBA3E (tubulin, alpha 3e)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR